Learn More
MilliporeSigma™ Supelco™ Solid Top Screw Cap with PTFE Liner
Known for providing the widest range of chemical compatibility and for being the most temperature tolerant of plastic closures
Supplier: MilliporeSigma™ Supelco™ 27174U
Description
- The color green easily identifies the PTFE liner system. F-217 foam-backed PTFE liners resist attack from virtually all chemicals at room temperature and provides odor and impact resilience for a tight seal with a low moisture transmission rate.
- F-217 has a low density foam core between two solid layers of LDPE (low density polyethylene).
- Some uses for these F-217/PTFE lined caps include analytical lab samples, high purity chemicals, strong acids solvents, environmental samples, pharmaceuticals, and diagnostic reagents.
Chemical Identifiers
| F217/PTFE | |
| Melamine Resin | |
| Solid Cap |
| Green | |
| 100 |
Specifications
| F217/PTFE | |
| Melamine Resin | |
| 20 mm | |
| Solid Cap |
| Green | |
| 100 | |
| 22 mL Vial |