missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
| Solanesol | |
| 42°C | |
| C45H74O | |
| 25 g | |
| CC(=CCCC(=CCCC(=CCCC(=CCCC(=CCCC(=CCCC(=CCCC(=CCCC(=CCO)C)C)C)C)C)C)C)C)C | |
| 631.086 | |
| CHEBI:26718 | |
| ≥93.0% (HPLC) |
| 13190-97-1 | |
| White-Yellow | |
| MFCD00070279 | |
| AFPLNGZPBSKHHQ-MEGGAXOGSA-N | |
| (2E,6E,10E,14E,18E,22E,26E,30E)-3,7,11,15,19,23,27,31,35-nonamethylhexatriaconta-2,6,10,14,18,22,26,30,34-nonaen-1-ol | |
| 5477212 | |
| 631.09 | |
| Crystalline Powder |
Chemical Identifiers
| 13190-97-1 | |
| 631.086 | |
| AFPLNGZPBSKHHQ-MEGGAXOGSA-N | |
| CHEBI:26718 | |
| CC(=CCCC(=CCCC(=CCCC(=CCCC(=CCCC(=CCCC(=CCCC(=CCCC(=CCO)C)C)C)C)C)C)C)C)C |
| C45H74O | |
| MFCD00070279 | |
| 5477212 | |
| (2E,6E,10E,14E,18E,22E,26E,30E)-3,7,11,15,19,23,27,31,35-nonamethylhexatriaconta-2,6,10,14,18,22,26,30,34-nonaen-1-ol |
Safety and Handling
RTECSNumber : MP5366666
TSCA : Yes
Recommended Storage : Refrigerator