accessibility menu, dialog, popup

UserName

CH3CH3CH3CH3CH3CH3CH3CH3CH3CH3OH(E)(E)(E)(E)(E)(E)(E)(E)

CAS RN 13190-97-1

IUPAC Name: (2E,6E,10E,14E,18E,22E,26E,30E)-3,7,11,15,19,23,27,31,35-nonamethylhexatriaconta-2,6,10,14,18,22,26,30,34-nonaen-1-ol
Synonyms: solanesol
Molecular Weight (g/mol): 631.09
Molecular Formula: C45H74O
InChi Key: AFPLNGZPBSKHHQ-MEGGAXOGSA-N
SMILES: CC(C)=CCC\C(C)=C\CC\C(C)=C\CC\C(C)=C\CC\C(C)=C\CC\C(C)=C\CC\C(C)=C\CC\C(C)=C\CC\C(C)=C\CO
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results