missing translation for 'onlineSavingsMsg'
Learn More

Solanesol 93.0+%, TCI America™

Catalog No. p-7135919
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S05255G 5 g
S052525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S05255G Supplier TCI America Supplier No. S05255G
Only null left

Chemical Identifiers

CAS 13190-97-1
Molecular Formula C45H74O
Molecular Weight (g/mol) 631.086
MDL Number MFCD00070279
InChI Key AFPLNGZPBSKHHQ-MEGGAXOGSA-N
PubChem CID 5477212
ChEBI CHEBI:26718
IUPAC Name (2E,6E,10E,14E,18E,22E,26E,30E)-3,7,11,15,19,23,27,31,35-nonamethylhexatriaconta-2,6,10,14,18,22,26,30,34-nonaen-1-ol
SMILES CC(=CCCC(=CCCC(=CCCC(=CCCC(=CCCC(=CCCC(=CCCC(=CCCC(=CCO)C)C)C)C)C)C)C)C)C

Specifications

Melting Point 42°C
Color White-Yellow
Quantity 5 g
Formula Weight 631.09
Percent Purity ≥93.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Solanesol
RTECSNumber MP5366666
TSCA Yes
Recommended Storage Refrigerator