missing translation for 'onlineSavingsMsg'
Learn More

Sodium Tungstate, Dihydrate, CP, Approx. 95%, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor S151625KG

 View more versions of this product

Catalog No. 18613963


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

10213-10-2
2.5 kg
Chemically Pure
1
∼95%
Poly Bottle

Chemical Identifiers

10213-10-2
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.