missing translation for 'onlineSavingsMsg'
Learn More

Sodium Tungstate, Dihydrate, CP, Approx. 95%, Spectrum™ Chemical

Catalog No. p-7166377
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
12 kg
2.5 kg
45 kg
500 g
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
Catalog No. Quantity Packaging
18613962 500 g Poly Bottle
18613963 2.5 kg Poly Bottle
18613964 12 kg Poly Pail
18613965 45 kg Fiber Drum
4 options

Catalog No. 18613962

Supplier: Spectrum Chemical Mfg Cor S1516500GM

Only null left

Chemical Identifiers

CAS 10213-10-2

Specifications

CAS Max % 1
Quantity 500 g
Percent Purity ∼95%
Grade Chemically Pure
Packaging Poly Bottle

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.