missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Trifluoroacetate 98.0+%, TCI America™
Supplier: TCI America T1336100G
Specifications
| Sodium Trifluoroacetate | |
| 207°C | |
| C2F3NaO2 | |
| 100 g | |
| sodium trifluoroacetate, trifluoroacetic acid sodium salt, acetic acid, trifluoro-, sodium salt, sodium perfluoroacetate, trifluoroacetate sodium, sodium 2,2,2-trifluoroacetate, trifluoroacetic acid sodium, trifluoroacetic acid,sodium salt, acetic acid, 2,2,2-trifluoro-, sodium salt 1:1, trifluoroacetic acid, sodium salt | |
| [Na+].[O-]C(=O)C(F)(F)F | |
| 136.01 | |
| 136.00 | |
| Crystalline Powder |
| 2923-18-4 | |
| White | |
| MFCD00013217 | |
| 2811 | |
| UYCAUPASBSROMS-UHFFFAOYSA-M | |
| sodium trifluoroacetate | |
| 517019 | |
| ≥98.0% (T) |
Chemical Identifiers
| 2923-18-4 | |
| 136.01 | |
| UYCAUPASBSROMS-UHFFFAOYSA-M | |
| 517019 | |
| [Na+].[O-]C(=O)C(F)(F)F |
| C2F3NaO2 | |
| MFCD00013217 | |
| sodium trifluoroacetate, trifluoroacetic acid sodium salt, acetic acid, trifluoro-, sodium salt, sodium perfluoroacetate, trifluoroacetate sodium, sodium 2,2,2-trifluoroacetate, trifluoroacetic acid sodium, trifluoroacetic acid,sodium salt, acetic acid, 2,2,2-trifluoro-, sodium salt 1:1, trifluoroacetic acid, sodium salt | |
| sodium trifluoroacetate |
Safety and Handling
EINECSNumber : (2)-3558
RTECSNumber : AK0250000
TSCA : Yes