missing translation for 'onlineSavingsMsg'
Learn More

Sodium Trifluoroacetate 98.0+%, TCI America™

Catalog No. p-7136720
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T133625G 25 g
T1336100G 100 g
T1336500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T133625G Supplier TCI America Supplier No. T133625G
Only null left

Chemical Identifiers

CAS 2923-18-4
Molecular Formula C2F3NaO2
Molecular Weight (g/mol) 136.01
MDL Number MFCD00013217
InChI Key UYCAUPASBSROMS-UHFFFAOYSA-M
Synonym sodium trifluoroacetate, trifluoroacetic acid sodium salt, acetic acid, trifluoro-, sodium salt, sodium perfluoroacetate, trifluoroacetate sodium, sodium 2,2,2-trifluoroacetate, trifluoroacetic acid sodium, trifluoroacetic acid,sodium salt, acetic acid, 2,2,2-trifluoro-, sodium salt 1:1, trifluoroacetic acid, sodium salt
PubChem CID 517019
IUPAC Name sodium trifluoroacetate
SMILES [Na+].[O-]C(=O)C(F)(F)F

Specifications

Melting Point 207°C
Color White
Quantity 25 g
UN Number 2811
Formula Weight 136.00
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Sodium Trifluoroacetate
EINECSNumber (2)-3558
RTECSNumber AK0250000
TSCA Yes