missing translation for 'onlineSavingsMsg'
Learn More

Sodium thiosulfate concentrate, For 1L standard solution, 0.1 M Na2S2O3 (0.1N), Honeywell Fluka™

Catalog No. p-7122109
Change view
Click to view available options
Quantity:
1 Pc.
6 x 1 Ea.
Catalog No. Quantity
6002675 1 Pc.
60046702 6 x 1 Ea.
2 options

Catalog No. 6002675

Supplier: Honeywell Chemicals 382001EA

Only null left

for 1L standard solution, 0.1 M Na2S2O3 (0.1N)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 10102-17-7 , 6132-02-1 , 7732-18-5
MDL Number MFCD00003499

Specifications

Quantity 1 Pc.
Molecular Formula Na2O3S2
Linear Formula Na2S2O3
UN Number NONH for all modes of transport
Synonym sodium thiosulfate, sodium thiosulphate, disodium thiosulfate, sodium thiosulfate anhydrous, hypo, sodiumthiosulfate, chlorine cure, chlorine control, declor-it, thiosulfuric acid, disodium salt
InChI Key AKHNMLFCWUSKQB-UHFFFAOYSA-L
SMILES [Na+].[Na+].[O-]S([S-])(=O)=O
IUPAC Name disodium;dioxido-oxo-sulfanylidene-$l^{6}-sulfane
Molecular Weight (g/mol) 158.10
PubChem CID 24477
Formula Weight 158.11g/mol
Packaging Ampoule
Chemical Name or Material Sodium thiosulfate concentrate
Show More Show Less