missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium thiosulfate concentrate, For 1L standard solution, 0.1 M Na2S2O3 (0.1N), Solstice
Description
- Leading the Industry For Over 65 Years
- Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
| Boiling Point | 100°C |
| CAS Min % | ≥20.0000% |
| Physical Form | Concentrate |
| CAS Max % | ≤40.0000%, ≤0.1000%, ≤59.9000% |
| Quantity | 6 x 1 Ea. |
| Packaging | Ampoule |
| Molecular Formula | H10Na2O8S2 |
| Linear Formula | Na2S2O3 |
| UN Number | NONH for all modes of transport |
| Synonym | sodium thiosulfate pentahydrate, ametox, sodium thiosulfate, pentahydrate, antichlor, tinver, disodium thiosulfate pentahydrate, unii-hx1032v43m, ccris 3952, thiosulfuric acid, disodium salt, pentahydrate, sodium hyposulfite pentahydrate |
| Show More |
Safety and Handling
| ShelfLife | 1260 days from date of manufacture |
| EINECSNumber | 231-791-2 |
| Recommended Storage | Room Temp |