missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium thiosulfate concentrate, For 1L standard solution, 0.01Â M Na2S2O3 (0.01N), Honeywell Fluka™
for 1L standard solution, 0.01Â M Na2S2O3 (0.01N)
$215.53 - $1124.21
Chemical Identifiers
| CAS | 7772-98-7 |
|---|---|
| Molecular Formula | Na2O3S2 |
| Molecular Weight (g/mol) | 158.10 |
| MDL Number | MFCD00003499 |
| InChI Key | AKHNMLFCWUSKQB-UHFFFAOYSA-L |
| Synonym | sodium thiosulfate, sodium thiosulphate, disodium thiosulfate, sodium thiosulfate anhydrous, hypo, sodiumthiosulfate, chlorine cure, chlorine control, declor-it, thiosulfuric acid, disodium salt |
| PubChem CID | 24477 |
| IUPAC Name | disodium;dioxido-oxo-sulfanylidene-$l^{6}-sulfane |
| SMILES | [Na+].[Na+].[O-]S([S-])(=O)=O |
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
| 7772-98-7 | |
| 158.10 | |
| AKHNMLFCWUSKQB-UHFFFAOYSA-L | |
| 24477 | |
| [Na+].[Na+].[O-]S([S-])(=O)=O |
| Na2O3S2 | |
| MFCD00003499 | |
| sodium thiosulfate, sodium thiosulphate, disodium thiosulfate, sodium thiosulfate anhydrous, hypo, sodiumthiosulfate, chlorine cure, chlorine control, declor-it, thiosulfuric acid, disodium salt | |
| disodium;dioxido-oxo-sulfanylidene-$l^{6}-sulfane |
Specifications
| 7772-98-7 | |
| Na2O3S2 | |
| MFCD00003499 | |
| sodium thiosulfate, sodium thiosulphate, disodium thiosulfate, sodium thiosulfate anhydrous, hypo, sodiumthiosulfate, chlorine cure, chlorine control, declor-it, thiosulfuric acid, disodium salt | |
| [Na+].[Na+].[O-]S([S-])(=O)=O | |
| 158.10 | |
| 158.11g/mol | |
| Sodium thiosulfate concentrate |
| 1 Pc. | |
| Na2S2O3 | |
| NONH for all modes of transport | |
| AKHNMLFCWUSKQB-UHFFFAOYSA-L | |
| disodium;dioxido-oxo-sulfanylidene-$l^{6}-sulfane | |
| 24477 | |
| Ampoule |
Safety and Handling
ShelfLife : 720 days from date of manufacture
Recommended Storage : Room Temp