missing translation for 'onlineSavingsMsg'
Learn More

Sodium Thiosulfate, Certified, 0.500N ±0.002N (0.5M), LabChem™

Catalog No. LC250802
Change view
Click to view available options
Quantity:
1 L
4 L
20 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC250802 1 L
LC250804 4 L
LC250805 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. LC250802 Supplier LabChem Supplier No. LC250802
Only null left

Chemical Identifiers

CAS 10102-17-7 , 7732-18-5 , 497-19-8
Molecular Formula H10Na2O8S2
Molecular Weight (g/mol) 248.17
InChI Key PODWXQQNRWNDGD-UHFFFAOYSA-L
Synonym sodium thiosulfate pentahydrate, ametox, sodium thiosulfate, pentahydrate, antichlor, tinver, disodium thiosulfate pentahydrate, unii-hx1032v43m, ccris 3952, thiosulfuric acid, disodium salt, pentahydrate, sodium hyposulfite pentahydrate
PubChem CID 61475
ChEBI CHEBI:32150
IUPAC Name disodium pentahydrate sulfanidesulfonate
SMILES O.O.O.O.O.[Na+].[Na+].[O-]S([S-])(=O)=O

Specifications

Color Colorless
CAS Max % 12.41, 87.57, 0.02
Physical Form Liquid
Quantity 1 L
Linear Formula Na2S2O3·5H2O
Solubility Information Soluble in water
Formula Weight 248.18
Grade Certified
Concentration or Composition (by Analyte or Components) 0.500N ±0.002N (0.5M)
Identification Passes Test
Packaging Poly Bottle
Decomposition Information Sulfur compounds; Carbon dioxide
Chemical Name or Material Sodium Thiosulfate
Show More Show Less
Hazard Statement GHS H Statement
Solution is not hazardous.
Precautionary Statement GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Recommended Storage Room Temperature