missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Thiosulfate, Certified, 0.1000N ±0.0005N (0.1M), LabChem™
Specifications
Specifications
| Color | Colorless |
| Physical Form | Liquid |
| CAS Max % | 2.48, 97.5, 0.02 |
| Quantity | 500 mL |
| Identification | Passes Test |
| Packaging | Poly Bottle |
| Decomposition Information | Sulfur compounds; Carbon dioxide |
| Linear Formula | Na2S2O3·5H2O |
| Solubility Information | Soluble in water |
| Formula Weight | 248.18 |
| Show More |
Safety and Handling
| Hazard Statement | GHS H Statement Solution is not hazardous. |
| Precautionary Statement | GHS P Statement If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes. If swallowed, get medical attention. |
| Recommended Storage | Room Temperature |