missing translation for 'onlineSavingsMsg'
Learn More

Sodium Thiosulfate, Anhydrous, 97%, Spectrum™ Chemical

$420.80 - $6362.70

Chemical Identifiers

CAS 7772-98-7
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18613954
View Documents
Spectrum Chemical Mfg Cor
S1498500GM
500 g Poly Bottle
Each for $420.80
Only null left
 
18613952
View Documents
Spectrum Chemical Mfg Cor
S149825KG
2.5 kg Poly Bottle
Each for $1,608.50
Only null left
 
18613951
View Documents
Spectrum Chemical Mfg Cor
S149812KG
12 kg Poly Pail
Each for $2,860.87
Only null left
 
18613953
View Documents
Spectrum Chemical Mfg Cor
S149845KGBL
45 kg Fiber Drum
Each for $6,362.70
Only null left
 
Description

Description

Specifications

Chemical Identifiers

7772-98-7

Specifications

7772-98-7
500 g
Reagent
1
97%
Poly Bottle
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.