missing translation for 'onlineSavingsMsg'
Learn More

Sodium Thiosulfate, Anhydrous, 97%, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor S149845KGBL

 View more versions of this product

Catalog No. 18613953


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

7772-98-7
45 kg
Reagent
1
97%
Fiber Drum

Chemical Identifiers

7772-98-7
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.