missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Thiosulfate, 0.100 N Aqueous Solution, APHA, Spectrum™ Chemical
Supplier: Spectrum Chemical Mfg Cor S5354LTGL
Specifications
| 540-72-7,7732-18-5,10102-17-7 | |
| CNNaS | |
| [Na+].[S-]C#N | |
| 81.07 | |
| Amber Glass Bottle |
| 4 L | |
| VGTPCRGMBIAPIM-UHFFFAOYSA-M | |
| sodium cyanosulfanide | |
| 0.100 N ± 0.0005 N |
Chemical Identifiers
| 540-72-7 | |
| 81.07 | |
| sodium cyanosulfanide |
| CNNaS | |
| VGTPCRGMBIAPIM-UHFFFAOYSA-M | |
| [Na+].[S-]C#N |
CAUTION: For laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.