missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Thiosulfate, 0.0375 N, Ricca Chemical
$95.09 - $173.97
Chemical Identifiers
| CAS | 10102-17-7 |
|---|---|
| Molecular Formula | H10Na2O8S2 |
| Molecular Weight (g/mol) | 248.17 |
| InChI Key | PODWXQQNRWNDGD-UHFFFAOYSA-L |
| Synonym | sodium thiosulfate pentahydrate, ametox, sodium thiosulfate, pentahydrate, antichlor, tinver, disodium thiosulfate pentahydrate, unii-hx1032v43m, ccris 3952, thiosulfuric acid, disodium salt, pentahydrate, sodium hyposulfite pentahydrate |
| PubChem CID | 61475 |
| ChEBI | CHEBI:32150 |
| IUPAC Name | disodium pentahydrate sulfanidesulfonate |
| SMILES | O.O.O.O.O.[Na+].[Na+].[O-]S([S-])(=O)=O |
Chemical Identifiers
| 10102-17-7 | |
| 248.17 | |
| sodium thiosulfate pentahydrate, ametox, sodium thiosulfate, pentahydrate, antichlor, tinver, disodium thiosulfate pentahydrate, unii-hx1032v43m, ccris 3952, thiosulfuric acid, disodium salt, pentahydrate, sodium hyposulfite pentahydrate | |
| CHEBI:32150 | |
| O.O.O.O.O.[Na+].[Na+].[O-]S([S-])(=O)=O |
| H10Na2O8S2 | |
| PODWXQQNRWNDGD-UHFFFAOYSA-L | |
| 61475 | |
| disodium pentahydrate sulfanidesulfonate |
Specifications
| Colorless | |
| 0.73,97.0,0.01 | |
| H10Na2O8S2 | |
| PODWXQQNRWNDGD-UHFFFAOYSA-L | |
| disodium pentahydrate sulfanidesulfonate | |
| 61475 | |
| Laboratory | |
| 1 L | |
| 0.0375Normal |
| 10102-17-7,7732-18-5,497-19-8 | |
| 0.76,100.0,0.01 | |
| sodium thiosulfate pentahydrate, ametox, sodium thiosulfate, pentahydrate, antichlor, tinver, disodium thiosulfate pentahydrate, unii-hx1032v43m, ccris 3952, thiosulfuric acid, disodium salt, pentahydrate, sodium hyposulfite pentahydrate | |
| O.O.O.O.O.[Na+].[Na+].[O-]S([S-])(=O)=O | |
| 248.17 | |
| CHEBI:32150 | |
| Natural Poly Bottle | |
| Liquid | |
| Sodium Thiosulfate |