missing translation for 'onlineSavingsMsg'
Learn More

Sodium Thiosulfate, 0.00500 N (N/200), Ricca Chemical

Oxidation-Reduction Titrant

$556.00 - $1375.13

Chemical Identifiers

CAS 10102-17-7
Molecular Formula H10Na2O8S2
Molecular Weight (g/mol) 248.17
InChI Key PODWXQQNRWNDGD-UHFFFAOYSA-L
Synonym sodium thiosulfate pentahydrate, ametox, sodium thiosulfate, pentahydrate, antichlor, tinver, disodium thiosulfate pentahydrate, unii-hx1032v43m, ccris 3952, thiosulfuric acid, disodium salt, pentahydrate, sodium hyposulfite pentahydrate
PubChem CID 61475
ChEBI CHEBI:32150
IUPAC Name disodium pentahydrate sulfanidesulfonate
SMILES O.O.O.O.O.[Na+].[Na+].[O-]S([S-])(=O)=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
78795-32
View Documents
Ricca Chemical Company
7879532
1 L Amber Glass Bottle
Each for $556.00
Only null left
 
7879-55
View Documents
Ricca Chemical Company
787955
20 L Cubitainer
Each for $1,375.13
Only null left
 
Description

Description

Specifications

Chemical Identifiers

10102-17-7
248.17
sodium thiosulfate pentahydrate, ametox, sodium thiosulfate, pentahydrate, antichlor, tinver, disodium thiosulfate pentahydrate, unii-hx1032v43m, ccris 3952, thiosulfuric acid, disodium salt, pentahydrate, sodium hyposulfite pentahydrate
CHEBI:32150
O.O.O.O.O.[Na+].[Na+].[O-]S([S-])(=O)=O
H10Na2O8S2
PODWXQQNRWNDGD-UHFFFAOYSA-L
61475
disodium pentahydrate sulfanidesulfonate

Specifications

Colorless
0.07,98.0,6.0E-4
H10Na2O8S2
PODWXQQNRWNDGD-UHFFFAOYSA-L
disodium pentahydrate sulfanidesulfonate
61475
Laboratory
Amber Glass Bottle
Liquid
10102-17-7,7732-18-5,497-19-8
0.07,100.0,6.0E-4
sodium thiosulfate pentahydrate, ametox, sodium thiosulfate, pentahydrate, antichlor, tinver, disodium thiosulfate pentahydrate, unii-hx1032v43m, ccris 3952, thiosulfuric acid, disodium salt, pentahydrate, sodium hyposulfite pentahydrate
O.O.O.O.O.[Na+].[Na+].[O-]S([S-])(=O)=O
248.17
CHEBI:32150
0.00500 Normal (N/200)
1 L
Sodium Thiosulfate
Videos
Safety and Handling

Safety and Handling

ShelfLife : 6 months

SDS
missing translation for 'documents'

missing translation for 'documents'