missing translation for 'onlineSavingsMsg'
Learn More

Sodium Thiosalicylate 95.0+%, TCI America™

Catalog No. T022025G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T022025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T022025G Supplier TCI America Supplier No. T022025G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 134-23-6
Molecular Formula C7H5NaO2S
Molecular Weight (g/mol) 176.165
MDL Number MFCD00070504
InChI Key HBAIZOJDXAXWHS-UHFFFAOYSA-M
Synonym Thiosalicylic Acid Sodium Salt, Sodium 2-Mercaptobenzoate, 2-Mercaptobenzoic Acid Sodium Salt
PubChem CID 23690429
ChEBI CHEBI:9182
IUPAC Name sodium;2-sulfanylbenzoate
SMILES C1=CC=C(C(=C1)C(=O)[O-])S.[Na+]

Specifications

Quantity 25 g
Formula Weight 176.16
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Sodium Thiosalicylate