accessibility menu, dialog, popup

UserName

CAS RN 134-23-6

IUPAC Name: sodium 2-sulfanylbenzoate
Synonyms: sodium thiosalicylate
Molecular Weight (g/mol): 176.16
Molecular Formula: C7H5NaO2S
InChi Key: HBAIZOJDXAXWHS-UHFFFAOYSA-M
SMILES: [Na+].[O-]C(=O)C1=CC=CC=C1S
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results