missing translation for 'onlineSavingsMsg'
Learn More

Sodium Thioglycolate 93.0+%, TCI America™

Supplier:  TCI America M0053100G

 View more versions of this product

Catalog No. M0053100G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Sodium Thioglycolate
White
C2H3NaO2S
100 g
GNBVPFITFYNRCN-UHFFFAOYSA-M
sodium;2-sulfanylacetate
23690444
114.09
Crystalline Powder
367-51-1
7.6 (5% in H2O)
MFCD00043386
sodium thioglycolate, sodium mercaptoacetate, mercaptoacetic acid sodium salt, mollescal sf, sodium 2-sulfanylacetate, thioglycolate sodium, sodium thioglycollate, mercaptoacetic acid, sodium salt, erhavit d, sodium 2-mercatoethanoate
C(C(=O)[O-])S.[Na+]
114.094
CHEBI:86481
≥93.0% (T)

Chemical Identifiers

367-51-1
114.094
GNBVPFITFYNRCN-UHFFFAOYSA-M
23690444
sodium;2-sulfanylacetate
C2H3NaO2S
MFCD00043386
sodium thioglycolate, sodium mercaptoacetate, mercaptoacetic acid sodium salt, mollescal sf, sodium 2-sulfanylacetate, thioglycolate sodium, sodium thioglycollate, mercaptoacetic acid, sodium salt, erhavit d, sodium 2-mercatoethanoate
CHEBI:86481
C(C(=O)[O-])S.[Na+]
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-1363

RTECSNumber : AI7700000

TSCA : Yes

Recommended Storage : Refrigerator

SDS
Documents

Documents

Promotions

Promotions