accessibility menu, dialog, popup

UserName

NaOOHS

CAS RN 367-51-1

IUPAC Name: sodium 2-sulfanylacetate
Synonyms: sodium thioglycolate
Molecular Weight (g/mol): 114.09
Molecular Formula: C2H3NaO2S
InChi Key: GNBVPFITFYNRCN-UHFFFAOYSA-M
SMILES: [Na+].[O-]C(=O)CS
Molecular Weight (g/mol) 
  • (5)
  • (12)
Percent Purity 
  • (3)
  • (2)
  • (3)
  • (9)
Quantity 
  • (2)
  • (5)
  • (4)
  • (1)
  • (1)
  • (4)
Physical Form 
  • (5)
  • (4)
  • (2)
Grade 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (8)
  • (2)
  • (2)
  • (2)
  • (3)
  • (10)
  • (5)
  • (12)
  • (3)
  • (2)
  • (3)
  • (9)
  • (2)
  • (5)
  • (4)
  • (1)
  • (1)
  • (4)
  • (5)
  • (4)
  • (2)
  • (2)
  • (2)