missing translation for 'onlineSavingsMsg'
Learn More

Sodium Thioglycolate 93.0+%, TCI America™

Catalog No. p-7131954
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M005325G 25 g
M0053100G 100 g
M0053500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. M005325G Supplier TCI America Supplier No. M005325G
Only null left

Chemical Identifiers

CAS 367-51-1
Molecular Formula C2H3NaO2S
Molecular Weight (g/mol) 114.094
MDL Number MFCD00043386
InChI Key GNBVPFITFYNRCN-UHFFFAOYSA-M
Synonym sodium thioglycolate, sodium mercaptoacetate, mercaptoacetic acid sodium salt, mollescal sf, sodium 2-sulfanylacetate, thioglycolate sodium, sodium thioglycollate, mercaptoacetic acid, sodium salt, erhavit d, sodium 2-mercatoethanoate
PubChem CID 23690444
ChEBI CHEBI:86481
IUPAC Name sodium;2-sulfanylacetate
SMILES C(C(=O)[O-])S.[Na+]

Specifications

Color White
pH 7.6 (5% in H2O)
Quantity 25 g
Formula Weight 114.09
Percent Purity ≥93.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Sodium Thioglycolate
EINECSNumber (2)-1363
RTECSNumber AI7700000
TSCA Yes
Recommended Storage Refrigerator