missing translation for 'onlineSavingsMsg'
Learn More

Sodium Thiocyanate, Crystal, ACS, 98%, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor S149512KG

 View more versions of this product

Catalog No. 18613903


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

540-72-7
12 kg
ACS
1
98%
Poly Pail

Chemical Identifiers

540-72-7
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.