Learn More
Sodium thiocyanate, 98+%, ACS reagent
CAS: 540-72-7 | CNNaS | 81.07 g/mol
Supplier: Thermo Scientific Chemicals 419675000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Sodium thiocyanate | |
| 540-72-7 | |
| Crystalline Powder, Crystals and/or Chunks | |
| 98% min. (Argentometry) | |
| NaSCN | |
| 01,1105 | |
| sodium thiocyanate, sodium rhodanate, sodium sulfocyanate, sodium rhodanide, thiocyanic acid, sodium salt, thiocyanate sodium, scyan, sodium isothiocyanate, nascn, haimased | |
| VGTPCRGMBIAPIM-UHFFFAOYSA-M | |
| sodium;thiocyanate | |
| 516871 | |
| 81.07 | |
| ACS Reagent |
| 287.0°C | |
| White | |
| 500 g | |
| CNNaS | |
| MFCD00011123 | |
| 15, 9480 | |
| Solubility in water: 1390g/L (21°C). Other solubilities: soluble in alcohol, freely soluble in acetone | |
| [Na+].[S-]C#N | |
| 81.07 | |
| CHEBI:30952 | |
| ≥98% | |
| Plastic bottle |
Chemical Identifiers
| 540-72-7 | |
| 81.07 | |
| VGTPCRGMBIAPIM-UHFFFAOYSA-M | |
| 516871 | |
| [Na+].[S-]C#N |
| CNNaS | |
| MFCD00011123 | |
| sodium thiocyanate, sodium rhodanate, sodium sulfocyanate, sodium rhodanide, thiocyanic acid, sodium salt, thiocyanate sodium, scyan, sodium isothiocyanate, nascn, haimased | |
| CHEBI:30952 |
Safety and Handling
GHS H Statement:
Harmful in contact with skin.
Harmful to aquatic life with long lasting effects.
Harmful if swallowed.
Harmful if inhaled.
Contact with acids liberates very toxic gas.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid release to the environment.
Keep container tightly closed.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
EINECSNumber : 208-754-4
RUO – Research Use Only