missing translation for 'onlineSavingsMsg'
Learn More

Sodium thiocyanate, 98+%, ACS reagent

Catalog No. p-3735381
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC419671000 100 g Plastic bottle
AC419675000 500 g Plastic bottle
AC419670025 2.5 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC419671000 Supplier Thermo Scientific Chemicals Supplier No. 419671000
Only null left

CAS: 540-72-7 | CNNaS | 81.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 540-72-7
Molecular Formula CNNaS
Molecular Weight (g/mol) 81.07
MDL Number MFCD00011123
InChI Key VGTPCRGMBIAPIM-UHFFFAOYSA-M
Synonym sodium thiocyanate, sodium rhodanate, sodium sulfocyanate, sodium rhodanide, thiocyanic acid, sodium salt, thiocyanate sodium, scyan, sodium isothiocyanate, nascn, haimased
PubChem CID 516871
ChEBI CHEBI:30952
SMILES [Na+].[S-]C#N

Specifications

Melting Point 287.0°C
Color White
Physical Form Crystalline Powder, Crystals and/or Chunks
Quantity 100 g
Assay Percent Range 98% min. (Argentometry)
Linear Formula NaSCN
Fieser 01,1105
Merck Index 15, 9480
Solubility Information Solubility in water: 1390g/L (21°C). Other solubilities: soluble in alcohol,freely soluble in acetone
IUPAC Name sodium;thiocyanate
Formula Weight 81.07
Percent Purity ≥98%
Grade ACS Reagent
Packaging Plastic bottle
Chemical Name or Material Sodium thiocyanate
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful in contact with skin.
Harmful to aquatic life with long lasting effects.
Harmful if swallowed.
Harmful if inhaled.
Contact with acids liberates very toxic gas.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid release to the environment.
Keep container tightly closed.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 208-754-4

RUO – Research Use Only