missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Tetraborate Decahydrate (Crystalline/Certified ACS), Fisher Chemical™
Supplier: Thermo Fisher Scientific S2483
Specifications
| Sodium Tetraborate Decahydrate | |
| White | |
| 9 | |
| 3 kg | |
| B4Na2O7 · 10 H2O | |
| Sodium Biborate, Sodium Pyroborate, Borax | |
| 99.5 to 105.0% | |
| Pass Test | |
| 1.7300g/cm³ |
| 75°C | |
| 1303-96-4 | |
| Powder/Solid | |
| 99.5 to 105.0 % | |
| MFCD00149193 | |
| 381.37 | |
| Certified ACS | |
| Poly Pail |
Chemical Identifiers
| 1303-96-4 | |
| B4Na2O7 · 10 H2O | |
| MFCD00149193 | |
| Sodium Biborate, Sodium Pyroborate, Borax |
Safety and Handling