missing translation for 'onlineSavingsMsg'
Learn More

Sodium Tetraborate Decahydrate (Crystalline/Certified ACS), Fisher Chemical™

Catalog No. p-33981
Change view
Click to view available options
Quantity:
500 g
3 kg
10 kg
Packaging:
Poly Bottle
Poly Pail
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S248500 500 g Poly Bottle
S2483 3 kg Poly Pail
S24810 10 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. S248500 Supplier Thermo Fisher Scientific Supplier No. S248 500
Only null left

Chemical Identifiers

CAS 1303-96-4
Molecular Formula B4Na2O7 · 10 H2O
MDL Number MFCD00149193
Synonym Sodium Biborate, Sodium Pyroborate, Borax

Specifications

Melting Point 75°C
Color White
pH 9
Physical Form Powder/Solid
Quantity 500 g
Assay Percent Range 99.5 to 105.0 %
Formula Weight 381.37
Percent Purity 99.5 to 105.0%
Grade Certified ACS
Identification Pass Test
Packaging Poly Bottle
Density 1.7300g/cm³
Chemical Name or Material Sodium Tetraborate Decahydrate
Show More Show Less