missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Tartrate, 10% (w/w), Ricca Chemical
$274.77 - $274.77
Chemical Identifiers
| CAS | 6106-24-7 |
|---|---|
| Molecular Formula | C4H8Na2O8 |
| Molecular Weight (g/mol) | 230.08 |
| MDL Number | MFCD00150035 |
| InChI Key | FGJLAJMGHXGFDE-UDMGOBQRNA-L |
| Synonym | unii-dia7c37aow, disodium tartrate dihydrate, disodium l-+-tartrate dihydrate, dia7c37aow, disodium +-tartrate dihydrate, sodium l-+-tartrate dihydrate, butanedioic acid, 2,3-dihydroxy-2r,3r-, disodium salt, dihydrate, disodium tartarate dihydrate, sodium tartrate nf, sodium tartrate dihydrate mi |
| PubChem CID | 131855972 |
| IUPAC Name | disodium (2R,3R)-2,3-dihydroxybutanedioate dihydrate |
| SMILES | O.O.[Na+].[Na+].O[C@H]([C@@H](O)C([O-])=O)C([O-])=O |
Chemical Identifiers
| 6106-24-7 | |
| 230.08 | |
| FGJLAJMGHXGFDE-UDMGOBQRNA-L | |
| 131855972 | |
| O.O.[Na+].[Na+].O[C@H]([C@@H](O)C([O-])=O)C([O-])=O |
| C4H8Na2O8 | |
| MFCD00150035 | |
| unii-dia7c37aow, disodium tartrate dihydrate, disodium l-+-tartrate dihydrate, dia7c37aow, disodium +-tartrate dihydrate, sodium l-+-tartrate dihydrate, butanedioic acid, 2,3-dihydroxy-2r,3r-, disodium salt, dihydrate, disodium tartarate dihydrate, sodium tartrate nf, sodium tartrate dihydrate mi | |
| disodium (2R,3R)-2,3-dihydroxybutanedioate dihydrate |
Specifications
| Colorless | |
| 88.0,9.81 | |
| C4H8Na2O8 | |
| unii-dia7c37aow, disodium tartrate dihydrate, disodium l-+-tartrate dihydrate, dia7c37aow, disodium +-tartrate dihydrate, sodium l-+-tartrate dihydrate, butanedioic acid, 2,3-dihydroxy-2r,3r-, disodium salt, dihydrate, disodium tartarate dihydrate, sodium tartrate nf, sodium tartrate dihydrate mi | |
| O.O.[Na+].[Na+].O[C@H]([C@@H](O)C([O-])=O)C([O-])=O | |
| 230.08 | |
| 10% | |
| Natural Poly Bottle | |
| 7 to 9 | |
| Liquid | |
| Sodium Tartrate |