missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium sulfite, 98%, for biochemistry
CAS: 7757-83-7 | Na2O3S | 126.04 g/mol
Supplier: Thermo Scientific Chemicals 447950010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Sodium sulfite | |
| 7757-83-7 | |
| 97.5% min. (Iodimetry) | |
| Na2SO3 | |
| sodium sulfite, disodium sulfite, sodium sulphite, sodium sulfite anhydrous, sulfurous acid, disodium salt, sulftech, natrium sulfurosum, sodiumsulfite, natriumsulfid, natriumsulfit | |
| GEHJYWRUCIMESM-UHFFFAOYSA-L | |
| disodium;sulfite | |
| 24437 | |
| 126.04 | |
| Biochemical | |
| 2.6300g/mL |
| >500.0°C | |
| 1 kg | |
| Na2O3S | |
| 15,8811 | |
| (1 M in water) Clear colorless | |
| [O-]S(=O)[O-].[Na+].[Na+] | |
| 126.04 | |
| CHEBI:86477 | |
| 98% | |
| 2.63 |
Chemical Identifiers
| 7757-83-7 | |
| 126.04 | |
| sodium sulfite, disodium sulfite, sodium sulphite, sodium sulfite anhydrous, sulfurous acid, disodium salt, sulftech, natrium sulfurosum, sodiumsulfite, natriumsulfid, natriumsulfit | |
| CHEBI:86477 | |
| [O-]S(=O)[O-].[Na+].[Na+] |
| Na2O3S | |
| GEHJYWRUCIMESM-UHFFFAOYSA-L | |
| 24437 | |
| disodium;sulfite |
Safety and Handling
EINECSNumber : 231-821-4
RUO – Research Use Only