missing translation for 'onlineSavingsMsg'
Learn More

Sodium sulfite, 98%, for biochemistry

Catalog No. p-4813667
Change view
Click to view available options
Quantity:
250 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC447952500 250 g
AC447950010 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC447952500 Supplier Thermo Scientific Chemicals Supplier No. 447952500
Only null left

CAS: 7757-83-7 | Na2O3S | 126.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7757-83-7
Molecular Formula Na2O3S
Molecular Weight (g/mol) 126.04
InChI Key GEHJYWRUCIMESM-UHFFFAOYSA-L
Synonym sodium sulfite, disodium sulfite, sodium sulphite, sodium sulfite anhydrous, sulfurous acid, disodium salt, sulftech, natrium sulfurosum, sodiumsulfite, natriumsulfid, natriumsulfit
PubChem CID 24437
ChEBI CHEBI:86477
IUPAC Name disodium;sulfite
SMILES [O-]S(=O)[O-].[Na+].[Na+]

Specifications

Melting Point >500.0°C
Quantity 250 g
Assay Percent Range 97.5% min. (Iodimetry)
Linear Formula Na2SO3
Merck Index 15,8811
Solubility Information (1 M in water) Clear colorless
Formula Weight 126.04
Percent Purity 98%
Grade Biochemical
Specific Gravity 2.63
Density 2.6300g/mL
Chemical Name or Material Sodium sulfite
Show More Show Less
EINECSNumber 231-821-4

RUO – Research Use Only