missing translation for 'onlineSavingsMsg'
Learn More

Sodium Sulfide, Solution No. 1 for Zinc, Certified, 3% (w/v), LabChem™

Catalog No. p-5388600
Change view
Click to view available options
Quantity:
125 mL
500 mL
1 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC249207 125 mL
LC249201 500 mL
LC249202 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. LC249207 Supplier LabChem Supplier No. LC249207
Only null left

Chemical Identifiers

CAS 1313-84-4 , 7732-18-5
Molecular Formula H18Na2O9S
Molecular Weight (g/mol) 240.17
MDL Number MFCD00149184
InChI Key ZGHLCBJZQLNUAZ-UHFFFAOYSA-N
IUPAC Name disodium nonahydrate sulfanediide
SMILES O.O.O.O.O.O.O.O.O.[Na+].[Na+].[S--]

Specifications

Color Colorless
CAS Max % 97, 3
Linear Formula Na2S·9H2O
Solubility Information Soluble in water
Formula Weight 240.17
Percent Purity 3% (w/v)
Grade Certified
Identification Passes Test
Odor Sulfide
Packaging Poly Bottle
Specific Gravity 1.006 ±0.002
Quantity 125 mL
Physical Form Liquid
Chemical Name or Material Sodium Sulfide, Solution No. 1 for Zinc
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye damage.
Very toxic to aquatic life.
Precautionary Statement GHS P Statement
Wear protective gloves, eye protection.
Wash exposed skin thoroughly after handling.
Avoid release to the environment.
If on skin: Wash with plenty of soap and water.
Take off contaminated clothing and wash before reuse.
If skin irritation occurs: Get medical advice/attention.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Collect spillage.
Dispose of contents/container to comply with local, state and federal regulations.
Recommended Storage Room Temperature