missing translation for 'onlineSavingsMsg'
Learn More

Sodium Sulfide, Solution No. 1 for Zinc, Certified, 3% (w/v), LabChem™

Supplier:  LabChem LC249201

 View more versions of this product

Catalog No. LC249201


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Sodium Sulfide
Solution No. 1 for Zinc
97,3
Na2S·9H2O
Soluble in water
O.O.O.O.O.O.O.O.O.[Na+].[Na+].[S--]
240.17
3% (w/v)
Passes Test
Poly Bottle
500 mL
Colorless
1313-84-4,7732-18-5
H18Na2O9S
MFCD00149184
ZGHLCBJZQLNUAZ-UHFFFAOYSA-N
disodium nonahydrate sulfanediide
240.17
Certified
Sulfide
1.006 ±0.002
Liquid

Chemical Identifiers

1313-84-4
240.17
ZGHLCBJZQLNUAZ-UHFFFAOYSA-N
O.O.O.O.O.O.O.O.O.[Na+].[Na+].[S--]
H18Na2O9S
MFCD00149184
disodium nonahydrate sulfanediide
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye damage.
Very toxic to aquatic life.

GHS P Statement
Wear protective gloves, eye protection.
Wash exposed skin thoroughly after handling.
Avoid release to the environment.
If on skin: Wash with plenty of soap and water.
Take off contaminated clothing and wash before reuse.
If skin irritation occurs: Get medical advice/attention.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Collect spillage.
Dispose of contents/container to comply with local, state and federal regulations.

Danger

Recommended Storage : Room Temperature

SDS
Documents

Documents

Promotions

Promotions