missing translation for 'onlineSavingsMsg'
Learn More

Sodium Sulfate, 22.2% (w/v), Ricca Chemical

Supplier:  Ricca Chemical Company 780032

Quantity 1 L
Packaging Natural Poly Bottle

 View more versions of this product

Catalog No. 780032

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

7757-82-6 , 7732-18-5
142.036
sodium sulfate, disodium sulfate, salt cake, thenardite, sodium sulfate anhydrous, sodium sulphate, sodium sulfate, anhydrous, sulfuric acid disodium salt, disodium sulphate, sodium sulfate 2:1
CHEBI:32149
[O-]S(=O)(=O)[O-].[Na+].[Na+]
Na2O4S
PMZURENOXWZQFD-UHFFFAOYSA-L
24436
disodium;sulfate

Specifications

Sodium Sulfate
80.0, 17.81
Na2O4S
PMZURENOXWZQFD-UHFFFAOYSA-L
disodium;sulfate
24436
Natural Poly Bottle
1 L
7757-82-6 , 7732-18-5
83.0, 18.54
sodium sulfate, disodium sulfate, salt cake, thenardite, sodium sulfate anhydrous, sodium sulphate, sodium sulfate, anhydrous, sulfuric acid disodium salt, disodium sulphate, sodium sulfate 2:1
[O-]S(=O)(=O)[O-].[Na+].[Na+]
142.036
CHEBI:32149
Aqueous Solution
22.2% (w/v)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions