missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Standard, 100 ppm Na, Ricca Chemical
$151.00 - $243.10
Chemical Identifiers
| CAS | 7647-14-5 |
|---|---|
| Molecular Formula | ClNa |
| Molecular Weight (g/mol) | 58.44 |
| MDL Number | MFCD00003477 |
| InChI Key | FAPWRFPIFSIZLT-UHFFFAOYSA-M |
| PubChem CID | 5234 |
| ChEBI | CHEBI:26710 |
| IUPAC Name | sodium chloride |
| SMILES | [Na+].[Cl-] |
Chemical Identifiers
| 7647-14-5 | |
| 58.44 | |
| FAPWRFPIFSIZLT-UHFFFAOYSA-M | |
| CHEBI:26710 | |
| [Na+].[Cl-] |
| ClNa | |
| MFCD00003477 | |
| 5234 | |
| sodium chloride |
Specifications
| Natural Poly Bottle | |
| 98.0,0.02 | |
| ClNa | |
| 100°C | |
| [Na+].[Cl-] | |
| 58.44 | |
| 5234 | |
| Sodium Standard, 1 mL = 0.1 mg Na |
| 7647-14-5,7732-18-5 | |
| 100.0,0.02 | |
| MFCD00003477 | |
| FAPWRFPIFSIZLT-UHFFFAOYSA-M | |
| sodium chloride | |
| 500 mL | |
| CHEBI:26710 |