missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium selenate, Purum p.a., ≥98.0% (T), Honeywell™
Purum p.a.,≥98.0% (T)
$3153.03 - $3153.03
Chemical Identifiers
| CAS | 13410-01-0 |
|---|---|
| Molecular Formula | Na2O4Se |
| Molecular Weight (g/mol) | 188.95 |
| MDL Number | MFCD00003490 |
| InChI Key | MHQOTKLEMKRJIR-UHFFFAOYSA-L |
| Synonym | sodium selenate, disodium selenate, natriumseleniat, selenic acid, disodium salt, caswell no. 791, natriumseleniat german, sel-tox sso2 and ss-20, unii-5dqp25600a, selenic acid h2seo4 , disodium salt, ccris 1259 |
| PubChem CID | 25960 |
| ChEBI | CHEBI:77775 |
| IUPAC Name | disodium;selenate |
| SMILES | [Na+].[Na+].[O-][Se]([O-])(=O)=O |
Description
- Products with a guarantee certificate and/or suitable for the stated analytical application.
Chemical Identifiers
| 13410-01-0 | |
| 188.95 | |
| MHQOTKLEMKRJIR-UHFFFAOYSA-L | |
| 25960 | |
| disodium;selenate |
| Na2O4Se | |
| MFCD00003490 | |
| sodium selenate, disodium selenate, natriumseleniat, selenic acid, disodium salt, caswell no. 791, natriumseleniat german, sel-tox sso2 and ss-20, unii-5dqp25600a, selenic acid h2seo4 , disodium salt, ccris 1259 | |
| CHEBI:77775 | |
| [Na+].[Na+].[O-][Se]([O-])(=O)=O |
Specifications
| 13410-01-0 | |
| Na2O4Se | |
| MFCD00003490 | |
| sodium selenate, disodium selenate, natriumseleniat, selenic acid, disodium salt, caswell no. 791, natriumseleniat german, sel-tox sso2 and ss-20, unii-5dqp25600a, selenic acid h2seo4 , disodium salt, ccris 1259 | |
| [Na+].[Na+].[O-][Se]([O-])(=O)=O | |
| 188.95 | |
| CHEBI:77775 | |
| ≥98% (T) | |
| Sodium selenate |
| 500 g | |
| Na2SeO4 | |
| UN2630 | |
| MHQOTKLEMKRJIR-UHFFFAOYSA-L | |
| disodium;selenate | |
| 25960 | |
| 188.94g/mol | |
| Poly bottle |
Safety and Handling
P261-P273-P301 + P310-P311-P501
RTECSNumber : VS6650000