accessibility menu, dialog, popup

UserName

NaNaOSeOOO

CAS RN 13410-01-0

IUPAC Name: disodium selenate
Synonyms: sodium selenate
Molecular Weight (g/mol): 188.95
Molecular Formula: Na2O4Se
InChi Key: MHQOTKLEMKRJIR-UHFFFAOYSA-L
SMILES: [Na+].[Na+].[O-][Se]([O-])(=O)=O
Grade 
  • (1)
  • (12)
Percent Purity 
  • (1)
  • (4)
  • (5)
  • (3)
Physical Form 
  • (1)
  • (3)
  • (5)
  • (4)
Quantity 
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (5)
  • (1)
  • (5)
  • (7)
  • (1)
  • (12)
  • (1)
  • (4)
  • (5)
  • (3)
  • (1)
  • (3)
  • (5)
  • (4)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)