missing translation for 'onlineSavingsMsg'
Learn More

Sodium Salicylate (Small Crystals, Free Flowing/USP), Fisher Chemical™

$770.71 - $770.71

Chemical Identifiers

CAS 54-21-7
Molecular Formula C7H5NaO3
Molecular Weight (g/mol) 160.104
MDL Number MFCD00002440
InChI Key ABBQHOQBGMUPJH-UHFFFAOYSA-M
Synonym sodium salicylate, sodium 2-hydroxybenzoate, salsonin, clin, enterosalicyl, enterosalil, entrosalyl, glutosalyl, kerasalicyl, magsalyl
PubChem CID 16760658
ChEBI CHEBI:9180
IUPAC Name sodium;2-hydroxybenzoate
SMILES C1=CC=C(C(=C1)C(=O)[O-])O.[Na+]
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
S396-500
SDS View Documents Promotion Details
Fisher Chemical
FLS396500
500 g Poly Bottle
Each for $770.71
Case of 4 Each for $3,082.84
Only null left
 
Description

Description

Specifications

Chemical Identifiers

54-21-7
160.104
ABBQHOQBGMUPJH-UHFFFAOYSA-M
16760658
sodium;2-hydroxybenzoate
C7H5NaO3
MFCD00002440
sodium salicylate, sodium 2-hydroxybenzoate, salsonin, clin, enterosalicyl, enterosalil, entrosalyl, glutosalyl, kerasalicyl, magsalyl
CHEBI:9180
C1=CC=C(C(=C1)C(=O)[O-])O.[Na+]

Specifications

54-21-7
White
Poly Bottle
2-HOC6H4COONa
500 g
Meets Requirements
C1=CC=C(C(=C1)C(=O)[O-])O.[Na+]
160.104
CHEBI:9180
99.5 to 100.5%
Solid
200°C
99.5 to 100.5 %
C7H5NaO3
MFCD00002440
sodium salicylate, sodium 2-hydroxybenzoate, salsonin, clin, enterosalicyl, enterosalil, entrosalyl, glutosalyl, kerasalicyl, magsalyl
ABBQHOQBGMUPJH-UHFFFAOYSA-M
sodium;2-hydroxybenzoate
16760658
160.1
USP
Sodium Salicylate
Videos
Safety and Handling

Safety and Handling

SDS
missing translation for 'documents'

missing translation for 'documents'