missing translation for 'onlineSavingsMsg'
Learn More

Sodium Potassium Tartrate, For Nitrogen (Ammonia), Certified, 30% (w/v), LabChem™

Catalog No. LC248301
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC248301 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. LC248301 Supplier LabChem Supplier No. LC248301

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6381-59-5 , 7732-18-5
Molecular Formula C4H12KNaO10
Molecular Weight (g/mol) 282.218
InChI Key VZOPRCCTKLAGPN-UHFFFAOYSA-L
Synonym potassium sodium tartrate tetrahydrate, sodium potassium tartrate tetrahydrate, monopotassium monosodium tartrate tetrahydrate, sodiumpotassiumtartrate,tetr, potassium sodium 2,3-dihydroxybutanedioate tetrahydrate, potassium sodium tetrahydrate tartrate, l +-tartaric acid potassium sodium salt, acmc-209nif, acmc-20aju8, ksc493o7j
PubChem CID 2724148
IUPAC Name potassium;sodium;2,3-dihydroxybutanedioate;tetrahydrate
SMILES C(C(C(=O)[O-])O)(C(=O)[O-])O.O.O.O.O.[Na+].[K+]

Specifications

Color Colorless
CAS Max % 70, 30
Linear Formula NaKC4H4O6·4H2O
Solubility Information Soluble in water
Formula Weight 282.23
Percent Purity 30% (w/v)
Grade Certified
Identification Passes Test
Packaging Poly Bottle
Suitability Passes Test
Decomposition Information Carbon monoxide; Carbon dioxide
Quantity 500 mL
Physical Form Liquid
Chemical Name or Material Sodium Potassium Tartrate, For Nitrogen (Ammonia)
Show More Show Less
Hazard Statement GHS H Statement
Solution is not hazardous.
Precautionary Statement GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Recommended Storage Refrigerate