missing translation for 'onlineSavingsMsg'
Learn More

Sodium Potassium Tartrate, 10% (w/v), pH 5.2, Ricca Chemical

$412.63 - $412.63

Chemical Identifiers

CAS 7732-18-5
Synonym dihydrogen oxide, dihydrogen monoxide
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
7540-32
SDS View Documents
Ricca Chemical Company
754032
1 L Natural Poly Bottle
Each for $412.63
Only null left
 
Description

Description

Specifications

Chemical Identifiers

7732-18-5
dihydrogen oxide, dihydrogen monoxide

Specifications

Colorless
89.0,8.91,0.02,0.02
dihydrogen oxide, dihydrogen monoxide
Laboratory
Aqueous Solution
1 L
10% (w/v)
7732-18-5,6381-59-5,9002-92-0,1310-73-2
93.0,9.27,0.02,0.02
10%
Natural Poly Bottle
5.2
Liquid
Sodium Potassium Tartrate, pH 5.2
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'