missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Picosulfate Monohydrate 98.0+%, TCI America™
Supplier: TCI America S09361G
Specifications
| Sodium Picosulfate Monohydrate | |
| 275°C | |
| C18H17NNa2O9S2 | |
| 1 g | |
| C1=CC=NC(=C1)C(C2=CC=C(C=C2)OS(=O)(=O)O)C3=CC=C(C=C3)OS(=O)(=O)O.O.[Na].[Na] | |
| 501.432 | |
| 481.40 | |
| Crystalline Powder |
| 1307301-38-7 | |
| White | |
| MFCD00867640 | |
| PUKNPYFKPBSERR-UHFFFAOYSA-N | |
| [4-[pyridin-2-yl-(4-sulfooxyphenyl)methyl]phenyl] hydrogen sulfate;sodium;hydrate | |
| 131871964 | |
| ≥98.0% (HPLC,T) |
Chemical Identifiers
| 1307301-38-7 | |
| 501.432 | |
| PUKNPYFKPBSERR-UHFFFAOYSA-N | |
| [4-[pyridin-2-yl-(4-sulfooxyphenyl)methyl]phenyl] hydrogen sulfate;sodium;hydrate |
| C18H17NNa2O9S2 | |
| MFCD00867640 | |
| 131871964 | |
| C1=CC=NC(=C1)C(C2=CC=C(C=C2)OS(=O)(=O)O)C3=CC=C(C=C3)OS(=O)(=O)O.O.[Na].[Na] |
Safety and Handling
RTECSNumber : SM8720000
TSCA : No