missing translation for 'onlineSavingsMsg'
Learn More

Sodium Picosulfate Monohydrate 98.0+%, TCI America™

Catalog No. p-7135996
Change view
Click to view available options
Quantity:
100 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S0936100MG 100 mg
S09361G 1 g
S09365G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. S0936100MG Supplier TCI America Supplier No. S0936100MG

Chemical Identifiers

CAS 1307301-38-7
Molecular Formula C18H17NNa2O9S2
Molecular Weight (g/mol) 501.432
MDL Number MFCD00867640
InChI Key PUKNPYFKPBSERR-UHFFFAOYSA-N
PubChem CID 131871964
IUPAC Name [4-[pyridin-2-yl-(4-sulfooxyphenyl)methyl]phenyl] hydrogen sulfate;sodium;hydrate
SMILES C1=CC=NC(=C1)C(C2=CC=C(C=C2)OS(=O)(=O)O)C3=CC=C(C=C3)OS(=O)(=O)O.O.[Na].[Na]

Specifications

Melting Point 275°C
Color White
Quantity 100 mg
Formula Weight 481.40
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Sodium Picosulfate Monohydrate
RTECSNumber SM8720000
TSCA No