accessibility menu, dialog, popup

UserName

NOSOOHOOSOOOHH2ONaNa

CAS RN 1307301-38-7

IUPAC Name: {4-[(pyridin-2-yl)[4-(sulfooxy)phenyl]methyl]phenyl}oxidanesulfonic acid hydrate disodium
Synonyms: (4-{pyridin-2-yl[4-(sulfooxy)phenyl]methyl}phenyl)oxidanesulfonic acid hydrate disodium
Molecular Weight (g/mol): 501.43
Molecular Formula: C18H17NNa2O9S2
InChi Key: PUKNPYFKPBSERR-UHFFFAOYSA-N
SMILES: O.[Na].[Na].OS(=O)(=O)OC1=CC=C(C=C1)C(C1=CC=C(OS(O)(=O)=O)C=C1)C1=CC=CC=N1
Percent Purity 
  • (3)
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)