missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Phosphate Tribasic Dodecahydrate, Purified (Trisodium Phosphate Dodecahydrate), Reagents
Sodium Phosphate Tribasic Dodecahydrate, Purified (Trisodium Phosphate Dodecahydrate), Chemically Pure, for general laboratory use. Manufactured in ISO 9001 facility.
Supplier: Labchem Inc C2324500500A5
Specifications
| Sodium Phosphate Tribasic Dodecahydrate, Purified (Trisodium Phosphate Dodecahydrate) | |
| 10101-89-0 | |
| 500 g | |
| Trisodium Phosphate Dodecahydrate | |
| O.O.O.O.O.O.O.O.O.O.O.O.[Na+].[Na+].[Na+].[O-]P([O-])([O-])=O | |
| 380.12 | |
| Poly Bottle |
| Colorless/White | |
| Solid | |
| H24Na3O16P | |
| ASTWEMOBIXQPPV-UHFFFAOYSA-K | |
| trisodium dodecahydrate phosphate | |
| Pure | |
| 1.6 |
Chemical Identifiers
| 10101-89-0 | |
| 380.12 | |
| Trisodium Phosphate Dodecahydrate | |
| O.O.O.O.O.O.O.O.O.O.O.O.[Na+].[Na+].[Na+].[O-]P([O-])([O-])=O |
| H24Na3O16P | |
| ASTWEMOBIXQPPV-UHFFFAOYSA-K | |
| trisodium dodecahydrate phosphate |