missing translation for 'onlineSavingsMsg'
Learn More

Sodium Phosphate Tribasic Dodecahydrate, Purified (Trisodium Phosphate Dodecahydrate), Reagents

Catalog No. p-200021019
Change view
Click to view available options
Quantity:
500 g
2.5 kg
10 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01336984 10 kg
01336983 2.5 kg
01336982 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01336984 Supplier Labchem Inc Supplier No. C232450010KG
Only null left

Sodium Phosphate Tribasic Dodecahydrate, Purified (Trisodium Phosphate Dodecahydrate), Chemically Pure, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 10101-89-0
Molecular Formula H24Na3O16P
Molecular Weight (g/mol) 380.12
InChI Key ASTWEMOBIXQPPV-UHFFFAOYSA-K
Synonym Trisodium Phosphate Dodecahydrate
IUPAC Name trisodium dodecahydrate phosphate
SMILES O.O.O.O.O.O.O.O.O.O.O.O.[Na+].[Na+].[Na+].[O-]P([O-])([O-])=O

Specifications

Color Colorless/White
Physical Form Solid
Quantity 10 kg
Grade Pure
Packaging Poly Pail
Specific Gravity 1.6
Chemical Name or Material Sodium Phosphate Tribasic Dodecahydrate, Purified (Trisodium Phosphate Dodecahydrate)