missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Phosphate Tribasic, Dodecahydrate, Crystal, Reagent, ACS, 98-102%, Spectrum™ Chemical
S1410, 10101-89-0, Na3PO4·12H2O
Supplier: Spectrum Chemical Mfg Cor S141025KGCS4
Description
Spectrum™ Chemical Sodium Phosphate Tribasic, Dodecahydrate, Crystal, Reagent, ACS is one of the sodium salts derived from phosphoric acid. Highly hygroscopic and water soluble it comes in the form of a white powder. Used commercially in powered product as an anticaking agent for those qualities. In an aqueous solution it is basic, with a pH value between 8.0 and 11.0. As an ACS grade Reagent, Spectrum Chemical manufactured compound is used as the quality standard against which other substances are grade and has met the toughest regulatory standards for quality and pureness.Specifications
| 10101-89-0 | |
| 4 x 2.5 kg | |
| ASTWEMOBIXQPPV-UHFFFAOYSA-K | |
| trisodium dodecahydrate phosphate | |
| 98 to 102% |
| 1 | |
| H24Na3O16P | |
| O.O.O.O.O.O.O.O.O.O.O.O.[Na+].[Na+].[Na+].[O-]P([O-])([O-])=O | |
| 380.12 | |
| Poly Bottle |
Chemical Identifiers
| 10101-89-0 | |
| 380.12 | |
| trisodium dodecahydrate phosphate |
| H24Na3O16P | |
| ASTWEMOBIXQPPV-UHFFFAOYSA-K | |
| O.O.O.O.O.O.O.O.O.O.O.O.[Na+].[Na+].[Na+].[O-]P([O-])([O-])=O |