missing translation for 'onlineSavingsMsg'
Learn More

Sodium Phosphate Tribasic, Dodecahydrate, Crystal, Reagent, ACS, 98-102%, Spectrum™ Chemical

Catalog No. p-7158256
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
6 x 500 g
12 kg
4 x 2.5 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18606722 125 g Poly Bottle
18606723 500 g Poly Bottle
18606725 2.5 kg Poly Bottle
18606724 6 x 500 g Poly Bottle
18606726 4 x 2.5 kg Poly Bottle
18606727 12 kg Poly Pail
18606728 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 18606722 Supplier Spectrum Chemical Mfg Cor Supplier No. S1410125GM
Only null left

S1410, 10101-89-0, Na3PO4·12H2O

Spectrum™ Chemical Sodium Phosphate Tribasic, Dodecahydrate, Crystal, Reagent, ACS is one of the sodium salts derived from phosphoric acid. Highly hygroscopic and water soluble it comes in the form of a white powder. Used commercially in powered product as an anticaking agent for those qualities. In an aqueous solution it is basic, with a pH value between 8.0 and 11.0. As an ACS grade Reagent, Spectrum Chemical manufactured compound is used as the quality standard against which other substances are grade and has met the toughest regulatory standards for quality and pureness.

Chemical Identifiers

CAS 10101-89-0
Molecular Formula H24Na3O16P
Molecular Weight (g/mol) 380.12
InChI Key ASTWEMOBIXQPPV-UHFFFAOYSA-K
IUPAC Name trisodium dodecahydrate phosphate
SMILES O.O.O.O.O.O.O.O.O.O.O.O.[Na+].[Na+].[Na+].[O-]P([O-])([O-])=O

Specifications

CAS Max % 1
Quantity 125 g
Percent Purity 98 to 102%
Grade ACS
Packaging Poly Bottle