missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Phosphate Monobasic, Monohydrate, Granular, BiotechGrade, 98-103%, Spectrum™ Chemical

S1396, 10049-21-5, NaH2PO4·H2O
$863.03 - $13467.70
Chemical Identifiers
CAS | 10049-21-5 |
---|---|
Molecular Formula | H5NaO5P |
Molecular Weight (g/mol) | 139.00 |
MDL Number | MFCD00149208 |
InChI Key | BBMHARZCALWXSL-UHFFFAOYSA-N |
IUPAC Name | sodium phosphoric acid hydrate |
SMILES | O.[Na+].OP(O)(O)=O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
18606706
|
Spectrum Chemical Mfg Cor
S1396500GM |
500 g | Poly Bottle |
Each for $863.03
|
|
|||||
18606708
|
Spectrum Chemical Mfg Cor
S139625KG |
2.5 kg | Poly Bottle |
Each for $2,883.03
|
|
|||||
18606707
|
Spectrum Chemical Mfg Cor
S1396500GMCS6 |
6 x 500 g | Poly Bottle |
Case of 6 Each for $4,499.17
|
|
|||||
18606709
|
Spectrum Chemical Mfg Cor
S139625KGCS4 |
4 x 2.5 kg | Poly Bottle |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | ||||||||||
18606710
|
Spectrum Chemical Mfg Cor
S139612KG |
12 kg | Poly Pail |
Each for $7,476.67
|
|
|||||
18606711
|
Spectrum Chemical Mfg Cor
S139650KGBL |
50 kg | Fiber Drum |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | ||||||||||
Description
Spectrum™ Chemical Sodium Phosphate Monobasic, Monohydrate, Granular, BiotechGrade is a reagent with very high buffering capacity widely used in molecular biology, biochemistry and chromatography. Spectrum offers highly pure reagents suitable for biochemical research and analysis that are absent of inhibitors.Chemical Identifiers
10049-21-5 | |
139.00 | |
BBMHARZCALWXSL-UHFFFAOYSA-N | |
O.[Na+].OP(O)(O)=O |
H5NaO5P | |
MFCD00149208 | |
sodium phosphoric acid hydrate |
Specifications
10049-21-5 | |
4.1 to 4.5 | |
H5NaO5P | |
BBMHARZCALWXSL-UHFFFAOYSA-N | |
sodium phosphoric acid hydrate | |
98 to 103% | |
Odorless | |
White granules |
1 | |
500 g | |
MFCD00149208 | |
O.[Na+].OP(O)(O)=O | |
139.00 | |
Biotech | |
Poly Bottle |