missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Phosphate Monobasic, Monohydrate, Granular, BiotechGrade, 98-103%, Spectrum™ Chemical
S1396, 10049-21-5, NaH2PO4·H2O
Supplier: Spectrum Chemical Mfg Cor S139625KG
Description
Spectrum™ Chemical Sodium Phosphate Monobasic, Monohydrate, Granular, BiotechGrade is a reagent with very high buffering capacity widely used in molecular biology, biochemistry and chromatography. Spectrum offers highly pure reagents suitable for biochemical research and analysis that are absent of inhibitors.Specifications
| 10049-21-5 | |
| 4.1 to 4.5 | |
| H5NaO5P | |
| BBMHARZCALWXSL-UHFFFAOYSA-N | |
| sodium phosphoric acid hydrate | |
| 98 to 103% | |
| Odorless | |
| White granules |
| 1 | |
| 2.5 kg | |
| MFCD00149208 | |
| O.[Na+].OP(O)(O)=O | |
| 139.00 | |
| Biotech | |
| Poly Bottle |
Chemical Identifiers
| 10049-21-5 | |
| 139.00 | |
| BBMHARZCALWXSL-UHFFFAOYSA-N | |
| O.[Na+].OP(O)(O)=O |
| H5NaO5P | |
| MFCD00149208 | |
| sodium phosphoric acid hydrate |