missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Phosphate Monobasic, ≥98%, MP Biomedicals
$117.73 - $339.75
Chemical Identifiers
| CAS | 7558-80-7 |
|---|---|
| Molecular Formula | H2NaO4P |
| Molecular Weight (g/mol) | 119.98 |
| MDL Number | MFCD00003527,MFCD00146206 |
| InChI Key | AJPJDKMHJJGVTQ-UHFFFAOYSA-M |
| Synonym | monosodium phosphate, sodium dihydrogen phosphate, sodium dihydrogenorthophosphate, sodium phosphate monobasic, sodium acid phosphate, sodium phosphate, monobasic, monosodium dihydrogen orthophosphate, sodium phosphate monobasic anhydrous, monosodium orthophosphate, monobasic sodium phosphate |
| PubChem CID | 23672064 |
| ChEBI | CHEBI:37585 |
| IUPAC Name | sodium dihydrogen phosphate |
| SMILES | [Na+].OP(O)([O-])=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
ICN19485083
|
MP Biomedicals Inc
0219485083 |
250 g |
Each for $117.73
|
|
|||||
|
ICN19485090
|
MP Biomedicals Inc
0219485090 |
500 g |
Each for $205.94
|
|
|||||
|
ICN19485091
|
MP Biomedicals Inc
0219485091 |
1 kg |
Each for $339.75
|
|
|||||
Chemical Identifiers
| 7558-80-7 | |
| 119.98 | |
| AJPJDKMHJJGVTQ-UHFFFAOYSA-M | |
| 23672064 | |
| sodium dihydrogen phosphate |
| H2NaO4P | |
| MFCD00003527,MFCD00146206 | |
| monosodium phosphate, sodium dihydrogen phosphate, sodium dihydrogenorthophosphate, sodium phosphate monobasic, sodium acid phosphate, sodium phosphate, monobasic, monosodium dihydrogen orthophosphate, sodium phosphate monobasic anhydrous, monosodium orthophosphate, monobasic sodium phosphate | |
| CHEBI:37585 | |
| [Na+].OP(O)([O-])=O |
Specifications
| 7558-80-7 | |
| ≥98% | |
| MFCD00003527,MFCD00146206 | |
| AJPJDKMHJJGVTQ-UHFFFAOYSA-M | |
| sodium dihydrogen phosphate | |
| 23672064 | |
| 120 |
| 250 g | |
| H2NaO4P | |
| monosodium phosphate, sodium dihydrogen phosphate, sodium dihydrogenorthophosphate, sodium phosphate monobasic, sodium acid phosphate, sodium phosphate, monobasic, monosodium dihydrogen orthophosphate, sodium phosphate monobasic anhydrous, monosodium orthophosphate, monobasic sodium phosphate | |
| [Na+].OP(O)([O-])=O | |
| 119.98 | |
| CHEBI:37585 | |
| Sodium Phosphate Monobasic, Anhydrous |
Safety and Handling
Recommended Storage : Room Temperature